missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Urea, ≥98%, Molecular Biology Reagent Grade, MP Biomedicals™

$63.89 - $725.85

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19485780
View Documents
MP Biomedicals Inc
0219485780
100 g
Each for $63.89
 
ICN19485790
View Documents
MP Biomedicals Inc
0219485790
500 g
Each for $140.00
 
ICN19485791
View Documents
MP Biomedicals Inc
0219485791
1 kg
Each for $214.78
 
ICN19485794
View Documents
MP Biomedicals Inc
0219485794
5 kg
Each for $725.85
 
Description

Description

Specifications

Chemical Identifiers

57-13-6
60.056
XSQUKJJJFZCRTK-UHFFFAOYSA-N
1176
urea
CH4N2O
MFCD00008022
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
CHEBI:48376
C(=O)(N)N

Specifications

57-13-6
1.335g/mL at 25°C (Lit.)
CH4N2O
100 g
XSQUKJJJFZCRTK-UHFFFAOYSA-N
urea
1176
60.06
Powder
White
≥98%
MFCD00008022
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
C(=O)(N)N
60.056
CHEBI:48376
Molecular Biology Reagent
Urea
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-315-5

Recommended Storage : Store at Room Temperature (15 to 30°C).

SDS
missing translation for 'documents'

missing translation for 'documents'