missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Urea, ≥98%, Molecular Biology Reagent Grade, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219485794

 View more versions of this product

Catalog No. ICN19485794


Explore available promotions

Description

Description

Specifications

Specifications

Urea
White
≥98%
MFCD00008022
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
C(=O)(N)N
60.056
CHEBI:48376
Molecular Biology Reagent
57-13-6
1.335g/mL at 25°C (Lit.)
CH4N2O
5 kg
XSQUKJJJFZCRTK-UHFFFAOYSA-N
urea
1176
60.06
Powder

Chemical Identifiers

57-13-6
60.056
XSQUKJJJFZCRTK-UHFFFAOYSA-N
1176
urea
CH4N2O
MFCD00008022
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
CHEBI:48376
C(=O)(N)N
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-315-5

Recommended Storage : Store at Room Temperature (15 to 30°C).

SDS
Documents

Documents

Promotions

Promotions