Learn More
Urea hydrogen peroxide, 1 g tablets, stabilized, contains 35 wt% H2O2, Thermo Scientific Chemicals
$319.32 - $1233.37
Chemical Identifiers
CAS | 124-43-6 |
---|---|
Molecular Formula | CH6N2O3 |
Molecular Weight (g/mol) | 94.07 |
InChI Key | AQLJVWUFPCUVLO-UHFFFAOYSA-N |
Synonym | urea hydrogen peroxide, carbamide peroxide, percarbamide, urea peroxide, urea dioxide, hydrogen peroxide urea, urea hydroperoxide, hydroperit, hydroperite, percarbamid |
PubChem CID | 31294 |
ChEBI | CHEBI:75178 |
IUPAC Name | hydrogen peroxide;urea |
SMILES | C(=O)(N)N.OO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
AC241020010
|
Thermo Scientific Chemicals
241020010 |
1 kg | Glass bottle |
Each for $319.32
|
|
||||
AC241020051
|
Thermo Scientific Chemicals
241020051 |
5 kg | Plastic drum |
Each for $1,233.37
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
124-43-6 | |
94.07 | |
urea hydrogen peroxide, carbamide peroxide, percarbamide, urea peroxide, urea dioxide, hydrogen peroxide urea, urea hydroperoxide, hydroperit, hydroperite, percarbamid | |
CHEBI:75178 | |
C(=O)(N)N.OO |
CH6N2O3 | |
AQLJVWUFPCUVLO-UHFFFAOYSA-N | |
31294 | |
hydrogen peroxide;urea |
Specifications
124-43-6,7722-88-5 | |
White | |
CH6N2O3 | |
1 kg | |
15,1784 | |
Solubility in water: 500g/L (20°C). Other solubilities: alcohol and ether decomposes the product to h2o2, and urea, soluble in ethylene glycol | |
C(=O)(N)N.OO | |
94.07 | |
31294 | |
94.07 | |
Tablets |
75.0°C to 85.0°C | |
Glass bottle | |
CO(NH2)2·H2O2 | |
16,379; 17,401 | |
urea hydrogen peroxide, carbamide peroxide, percarbamide, urea peroxide, urea dioxide, hydrogen peroxide urea, urea hydroperoxide, hydroperit, hydroperite, percarbamid | |
AQLJVWUFPCUVLO-UHFFFAOYSA-N | |
hydrogen peroxide;urea | |
1% sodium pyrophosphate | |
CHEBI:75178 | |
≥33.5% (H2O2) | |
Urea hydrogen peroxide |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 204-701-4
RUO – Research Use Only