missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Urethane, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 325542500

 View more versions of this product

Catalog No. AC325542500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Urethane
51-79-6
White
92°C
96% min. (GC)
C3H7NO2
MFCD00007966
03,22
15,10059
Solubility in water: 200g/100 mL (20°C). Other solubilities: soluble in ethanol (125g/100 mL),chloroform,(111g/100 mL),ether (66.67g/100 mL),glycerol,(40g/100 mL),,slightly soluble in olive oil (3g/100 mL)
CCOC(=O)N
89.09
CHEBI:17967
97%
97%
47.0°C to 51.0°C
182.0°C to 184.0°C
Authentic
Plastic bottle
H2NCO2C2H5
250 g
01,127
urethane, urethan, ethylurethane, carbamic acid ethyl ester, ethyl urethane, carbamic acid, ethyl ester, ethylcarbamate, pracarbamine, leucethane, pracarbamin
JOYRKODLDBILNP-UHFFFAOYSA-N
ethyl carbamate
5641
89.09
Crystalline Powder or Crystals

Chemical Identifiers

51-79-6
89.09
JOYRKODLDBILNP-UHFFFAOYSA-N
5641
ethyl carbamate
C3H7NO2
MFCD00007966
urethane, urethan, ethylurethane, carbamic acid ethyl ester, ethyl urethane, carbamic acid, ethyl ester, ethylcarbamate, pracarbamine, leucethane, pracarbamin
CHEBI:17967
CCOC(=O)N
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause cancer.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Obtain special instructions before us

GHS Signal Word: Danger

EINECSNumber : 200-123-1

RTECSNumber : FA8400000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only