Learn More
Urethane, 97%, Thermo Scientific Chemicals
$52.31 - $169.23
Chemical Identifiers
CAS | 51-79-6 |
---|---|
Molecular Formula | C3H7NO2 |
Molecular Weight (g/mol) | 89.09 |
MDL Number | MFCD00007966 |
InChI Key | JOYRKODLDBILNP-UHFFFAOYSA-N |
Synonym | urethane, urethan, ethylurethane, carbamic acid ethyl ester, ethyl urethane, carbamic acid, ethyl ester, ethylcarbamate, pracarbamine, leucethane, pracarbamin |
PubChem CID | 5641 |
ChEBI | CHEBI:17967 |
IUPAC Name | ethyl carbamate |
SMILES | CCOC(=O)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC325540500
|
Thermo Scientific Chemicals
325540500 |
50 g | Plastic bottle |
Each for $52.31
|
|
||||
AC325542500
|
Thermo Scientific Chemicals
325542500 |
250 g | Plastic bottle |
Each for $169.23
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
51-79-6 | |
89.09 | |
JOYRKODLDBILNP-UHFFFAOYSA-N | |
5641 | |
ethyl carbamate |
C3H7NO2 | |
MFCD00007966 | |
urethane, urethan, ethylurethane, carbamic acid ethyl ester, ethyl urethane, carbamic acid, ethyl ester, ethylcarbamate, pracarbamine, leucethane, pracarbamin | |
CHEBI:17967 | |
CCOC(=O)N |
Specifications
51-79-6 | |
White | |
92°C | |
96% min. (GC) | |
C3H7NO2 | |
MFCD00007966 | |
03,22 | |
15,10059 | |
Solubility in water: 200g/100 mL (20°C). Other solubilities: soluble in ethanol (125g/100 mL),chloroform,(111g/100 mL),ether (66.67g/100 mL),glycerol,(40g/100 mL),,slightly soluble in olive oil (3g/100 mL) | |
CCOC(=O)N | |
89.09 | |
CHEBI:17967 | |
97% | |
Urethane, 97% |
47.0°C to 51.0°C | |
182.0°C to 184.0°C | |
Authentic | |
Plastic bottle | |
H2NCO2C2H5 | |
50 g | |
01,127 | |
urethane, urethan, ethylurethane, carbamic acid ethyl ester, ethyl urethane, carbamic acid, ethyl ester, ethylcarbamate, pracarbamine, leucethane, pracarbamin | |
JOYRKODLDBILNP-UHFFFAOYSA-N | |
ethyl carbamate | |
5641 | |
89.09 | |
Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause cancer.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Obtain special instructions before us
GHS Signal Word: Danger
EINECSNumber : 200-123-1
RTECSNumber : FA8400000
TSCA : TSCA
RUO – Research Use Only