Greiner Bio-One Urine Cup with Integrated Transfer Device, 100 mL
The Urine Cup with Integrated Transfer Device allows you to collect and transport urine samples safely to the necessary destination.
Supplier: Greiner Bio-One 724322
Description
- Recessed transfer device reduces risk of needlestick injuries.
- Bulk-packaged to reduce waste and associated costs.
- Perforated label acts as an integrity seal, visually indicating if the product has been opened prior to use.
- Optimized threading for easy open and close, with a stabilizing flange that prevents over-threading.
- Leak-proof with manual transport and drop proof (1.2 meters).
Specifications
Urine Collection Cup | |
Screw Cap | |
Yes | |
Non-sterile | |
100 mL |
Yellow | |
Single-use | |
Polypropylene | |
Yes |