Honeywell Uvex Genesis™ Replacement Lens
Economical and easy lens replacement allows for multiple lens tints, extending life of eyewear, increasing value and lowering cost of ownership. Lens meets impact resistance standards and provide superior optics and unobstructed peripheral vision.
Supplier: Honeywell S6907HS
Description
- Lens meets impact resistance standards and provide superior optics and unobstructed peripheral vision.
- Superior Hydroshield anti-fog coating keeps glasses consistently fog-free for 90X longer than the majority of other anti-fog products and 2X greater scratch resistance than our next best anti-fog coating
Specifications
Anti-fog | |
Shade 3 | |
Polycarbonate | |
Polycarbonate |
Hydroshield Anti-Fog | |
Replacement Lens | |
Reusable | |
UV400 |