UVP Large Chromato-Vue™ Viewing Cabinets
Cabinets designed for long life and rugged use in a wide range of applications
$5465.44 - $6077.79
Catalog Number | Mfr. No. | Model | Product Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Product Type | Price | Quantity | ||||
UVP95002201
|
UVP
95002201 |
C-71 | 365nm viewing cabinet |
Each for $5,772.88
|
|
||||
UVP95002004
|
UVP
95002004 |
C-70G | 254/365nm viewing cabinet |
Each for $5,465.44
|
|
||||
UVP95025301
|
UVP
95025301 |
C-75 | 254/365nm viewing cabinet |
Each for $6,077.79
|
|
||||
Description
- Durable lightweight plastic made
- View port combines a comfortable soft rubber and a contrast control filter
- Removable bottom panel
- Fitted with ultraviolet light sources
Model C-75:
- Illumination is from 15-watt shortwave and longwave tubes positioned on two sides of the cabinet interior
- internal white light is operated by the rocker switch located on top of the cabinet
- Includes two each 15-watt shortwave and longwave ultraviolet tubes
- Includes four 15-watt longwave ultraviolet tubes
Specifications
Viewing materials |