UVP Large Chromato-Vue™ Viewing Cabinets
Cabinets designed for long life and rugged use in a wide range of applications
Supplier:  UVP 95002004
 View more versions of this product
Catalog No. UVP95002004
Explore available promotions
Description
Description
  • Durable lightweight plastic made
  • View port combines a comfortable soft rubber and a contrast control filter
  • Removable bottom panel
  • Fitted with ultraviolet light sources
Model C-75:
  • Illumination is from 15-watt shortwave and longwave tubes positioned on two sides of the cabinet interior
  • internal white light is operated by the rocker switch located on top of the cabinet
Model C-70G:
  • Includes two each 15-watt shortwave and longwave ultraviolet tubes
Model C-71:
  • Includes four 15-watt longwave ultraviolet tubes
Specifications
Specifications
C-70G
Viewing materials
254/365nm viewing cabinet