UVP White Light Replacement Tubes
Supplier:  UVP 34005601
 View more versions of this product
Catalog No. UVP34005601
Explore available promotions
Description
Description
Specifications
Specifications
Replacement Tube
Models TLW-20, TMW-20, TW-26 transilluminator
11.33 in.
1 Ea.
8 W
8 W cool white tube
28.7 cm