missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

UVP White Light Replacement Tubes

$34.37 - $98.09

Specifications

Product Type Replacement Tube
Packaging Quantity 1 Ea.
View More Specs

Products 5
Catalog Number Mfr. No. For Use With (Equipment) Wattage Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Wattage Price Quantity  
UVP34005602
View Documents
UVP
34005602
Model MRL-58 multiple-ray lamp 8 W
Each for $76.63
 
UVP34005901
View Documents
UVP
34005901
Model TW-43 white light transilluminator 14 W
Each for $98.09
 
UVP34005601
View Documents
UVP
34005601
Models TLW-20, TMW-20, TW-26 transilluminator 8 W
Each for $72.30
 
UVP34006601
View Documents
UVP
34006601
14 Type EL series UV lamp 4 W
Each for $45.82
 
UVP34006301
View Documents
UVP
34006301
16 Type EL series UV lamp and high performance transilluminator 6 W
Each for $34.37
 
Description

Description

Specifications

Specifications

Replacement Tube
1 Ea.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'