UVP White Light Replacement Tubes
Supplier:  UVP 34005901
 View more versions of this product
Catalog No. UVP34005901
Explore available promotions
Description
Description
Specifications
Specifications
14 W
4.25 in.
14 W cool white tube
16 in.
Replacement Tube
Model TW-43 white light transilluminator
10.8 cm
1 Ea.
40.6 cm