BD Vacutainer™ UltraTouch™ Push Button Blood Collection Set
Engineered to minimize patient discomfort during blood collection
Supplier:  BD 367393
 View more versions of this product
Catalog No. B367393
Explore available promotions
Description
Description
  • Singe-handed, in-vein safety activation instantly retracts needle after use
  • PentaPoint™ bevel requires 32% less penetration force
  • Ultra-thin RightGauge™ cannula allows the use of a smaller gauge suitable for more veins, without compromising filling times or sample quality
Specifications
Specifications
21 G
0.75 in.
7 in.