BD Vacutainer™ UltraTouch™ Push Button Blood Collection Set
Engineered to minimize patient discomfort during blood collection
$237.48 - $1187.77
Catalog Number | Mfr. No. | Needle Gauge | Length (English) Needle | Length (English) Tube | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Needle Gauge | Length (English) Needle | Length (English) Tube | Price | Quantity | |||
B367363
|
BD
367363 |
25 G | 0.75 in. | 12 in. |
Pack of 50 Each for $237.48
Case of 4 Pack for $949.92
|
|
|||
B367364
|
BD
367364 |
23 G | 0.75 in. | 12 in. |
Pack of 50 Each for $237.48
Case of 4 Pack for $949.92
|
|
|||
B367365
|
BD
367365 |
21 G | 0.75 in. | 12 in. |
Pack of 50 Each for $237.48
Case of 4 Pack for $949.92
|
|
|||
B367393
|
BD
367393 |
21 G | 0.75 in. | 7 in. |
Pack of 50 Each for $296.95
Case of 4 Pack for $1,187.80
|
|
|||
B367392
|
BD
367392 |
23 G | 0.75 in. | 7 in. |
Case of 200 Each for $1,187.77
|
|
|||
Description
- Singe-handed, in-vein safety activation instantly retracts needle after use
- PentaPoint™ bevel requires 32% less penetration force
- Ultra-thin RightGauge™ cannula allows the use of a smaller gauge suitable for more veins, without compromising filling times or sample quality
Specifications
0.75 in. |