missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD Vacutainer™ Urine Collection Kits: Combination
Reduce the need to retest, relabel, and/or recollect specimens
Supplier: BD 364956
Description
- Closed systems that benefit healthcare workers by reducing their need to come into contact with potentially hazardous specimens
- Designed to provide safety and satisfaction, as well as consistent, reliable results to ensure proper patient outcomes
Specifications
Combination Urine Specimen Collection Kit | |
Sterile | |
Integrated transfer device, plastic C and S preservative, evacuated Plastic conical bottom tube, sterile, screw-cap cup, castile soap towelettes | |
0.27 oz. |
Plastic | |
For microbiology urine testing | |
Room temperature | |
8 mL |