missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD Vacutainer™ Urine Collection Kits: Combination
Reduce the need to retest, relabel, and/or recollect specimens
$284.46 - $385.80
Specifications
Product Type | Combination Urine Specimen Collection Kit |
---|---|
Material | Plastic |
Sterility | Sterile |
For Use With (Application) | For microbiology urine testing |
Storage Requirements | Room temperature |
Catalog Number | Mfr. No. | Volume (English) | Volume (Metric) | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (English) | Volume (Metric) | Includes | Price | Quantity | ||||
02-683-193
|
BD
364956 |
0.27 oz. | 8 mL | Integrated transfer device, plastic C and S preservative, evacuated Plastic conical bottom tube, sterile, screw-cap cup, castile soap towelettes |
Case of 50 Each for $284.46
|
|
||||
02-683-194
|
BD
364957 |
0.135 oz., 0.27 oz. | 4 mL, 8 mL | Plastic C and S preservative, evacuated Plastic conical bottom tube, sterile, integrated transfer device, castile soap towelettes, 4 mL 13 x 75 mm Plastic C and S preservative tube, soap towelette and cup |
Case of 50 Each for $385.80
|
|
||||
Description
- Closed systems that benefit healthcare workers by reducing their need to come into contact with potentially hazardous specimens
- Designed to provide safety and satisfaction, as well as consistent, reliable results to ensure proper patient outcomes
Specifications
Combination Urine Specimen Collection Kit | |
Sterile | |
Room temperature |
Plastic | |
For microbiology urine testing |