Synthware™ Vacuum and Inert Gas Manifold with Hollow Glass Stopcocks, Front Left/Right, Rear/Left Connection
No retention clip needed. Hollow stopcocks held in place by vacuum.
Supplier: Synthware™ M421004
Description
High vacuum hollow glass stopcocks precision ground with corresponding numbers on barrel and plug. Lower vacuum chambers on stopcock are evacuated with a separate hole in the plug in order to hold the plug in the stopcock barrels.
Hose connection : Back Beam-Left Only, Front Beam-Both Ends
Specifications
Front Left and Right; Rear Left Hose Connection | |
Organic Synthesis | |
Borosilicate Glass | |
375 mm |
Vacuum/Inert Gas Manifold | |
Hollow Glass Stopcocks | |
4 |