Fischer Technical Company™ Cast Iron High Vacuum Pump Plate DFS Item
Cast Iron High Vacuum Pump Plate is high quality for years of classroom use.
Supplier:  Fischer Technical Company™ GA100
 View more versions of this product
Catalog No. S41330-1
Explore available promotions
Description
Description
  • Made of steel with rubber gasket
  • Large enough to accommodate bell jars up to 9 in. (22.5 cm) in diameter
  • Nickel-plated, 0.5 in. (1.25 cm) steel plate provides extra strength and is ground flat for a good seal
  • Complete with a barbed hose connection, vacuum release valve and rubber gasket to provide a tight seal
  • Replacement Gaskets Available
Specifications
Specifications
Vacuum Plate
Classroom Use
Barbed Hose Connection, Vacuum Release Valve, Rubber Gasket