missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Valerophenone, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 140800250

 View more versions of this product

Catalog No. AC140800250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Valerophenone
1009-14-9
Green-Yellow to Yellow
105.0°C to 107.0°C (5.0 mmHg)
Authentic
Glass bottle
1.5143 to 1.5163
MFCD00009480
07,327
valerophenone, butyl phenyl ketone, pentanophenone, 1-phenyl-1-pentanone, 1-pentanone, 1-phenyl, unii-f27q043nt1, n-butyl phenyl ketone, 1-benzoylbutane, valerophenone, 8ci
CCCCC(=O)C1=CC=CC=C1
162.23
CHEBI:36812
98%
98%
-9.0°C
0.9800g/mL
102°C
97.5% min. (GC)
C11H14O
C6H5CO(CH2)3CH3
25 g
0.98
XKGLSKVNOSHTAD-UHFFFAOYSA-N
1-phenylpentan-1-one
66093
162.23
Liquid

Chemical Identifiers

1009-14-9
162.23
XKGLSKVNOSHTAD-UHFFFAOYSA-N
66093
1-phenylpentan-1-one
C11H14O
MFCD00009480
valerophenone, butyl phenyl ketone, pentanophenone, 1-phenyl-1-pentanone, 1-pentanone, 1-phenyl, unii-f27q043nt1, n-butyl phenyl ketone, 1-benzoylbutane, valerophenone, 8ci
CHEBI:36812
CCCCC(=O)C1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 213-767-3

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only