missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Valerophenone, 98%, Thermo Scientific Chemicals
$28.92 - $86.51
Chemical Identifiers
CAS | 1009-14-9 |
---|---|
Molecular Formula | C11H14O |
Molecular Weight (g/mol) | 162.23 |
MDL Number | MFCD00009480 |
InChI Key | XKGLSKVNOSHTAD-UHFFFAOYSA-N |
Synonym | valerophenone, butyl phenyl ketone, pentanophenone, 1-phenyl-1-pentanone, 1-pentanone, 1-phenyl, unii-f27q043nt1, n-butyl phenyl ketone, 1-benzoylbutane, valerophenone, 8ci |
PubChem CID | 66093 |
ChEBI | CHEBI:36812 |
IUPAC Name | 1-phenylpentan-1-one |
SMILES | CCCCC(=O)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC140800050
|
Thermo Scientific Chemicals
140800050 |
5 g | Glass bottle |
Each for $28.92
|
|
||||
AC140800250
|
Thermo Scientific Chemicals
140800250 |
25 g | Glass bottle |
Each for $86.51
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1009-14-9 | |
162.23 | |
XKGLSKVNOSHTAD-UHFFFAOYSA-N | |
66093 | |
1-phenylpentan-1-one |
C11H14O | |
MFCD00009480 | |
valerophenone, butyl phenyl ketone, pentanophenone, 1-phenyl-1-pentanone, 1-pentanone, 1-phenyl, unii-f27q043nt1, n-butyl phenyl ketone, 1-benzoylbutane, valerophenone, 8ci | |
CHEBI:36812 | |
CCCCC(=O)C1=CC=CC=C1 |
Specifications
1009-14-9 | |
Green-Yellow to Yellow | |
105.0°C to 107.0°C (5.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5143 to 1.5163 | |
MFCD00009480 | |
07,327 | |
valerophenone, butyl phenyl ketone, pentanophenone, 1-phenyl-1-pentanone, 1-pentanone, 1-phenyl, unii-f27q043nt1, n-butyl phenyl ketone, 1-benzoylbutane, valerophenone, 8ci | |
CCCCC(=O)C1=CC=CC=C1 | |
162.23 | |
CHEBI:36812 | |
98% | |
Valerophenone, 98% |
-9.0°C | |
0.9800g/mL | |
102°C | |
97.5% min. (GC) | |
C11H14O | |
C6H5CO(CH2)3CH3 | |
5 g | |
0.98 | |
XKGLSKVNOSHTAD-UHFFFAOYSA-N | |
1-phenylpentan-1-one | |
66093 | |
162.23 | |
Liquid |
Safety and Handling
EINECSNumber : 213-767-3
TSCA : TSCA
RUO – Research Use Only