DWK Life Sciences Kimble™ ValueWare™ Volumetric Flasks, Class A
Calibrated “To Contain.”
Supplier:  DWK Life Sciences 5801425
 View more versions of this product
Catalog No. S00121
Description
Description
  • Kimax™ KG-33 glass Single graduation ring blasted on the neck
  • With plug stopcock ground glass stopper and marking spot
Compliance
Glass meets Type 1, Class A, ASTM™ E-438 standards. Designed to ASTM Specification E288, Class A unserialized requirements.
Specifications
Specifications
Calibrated To Contain
25 mL
9
Glass
Glass