DWK Life Sciences Kimble™ ValueWare™ Volumetric Flasks, Class A
Calibrated “To Contain.”
$51.64 - $797.38
Specifications
Calibration | Calibrated To Contain |
---|---|
Closure Material | Glass |
Material | Glass |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
S00120
|
DWK Life Sciences
5801410 |
10 mL |
Case of 12 Each for $745.74
|
|
|||||
S00121
|
DWK Life Sciences
5801425 |
25 mL |
Pack of 6 Each for $453.21
Case of 2 Pack for $906.42
|
|
|||||
S00122
|
DWK Life Sciences
5801450 |
50 mL |
Pack of 6 Each for $482.97
Case of 2 Pack for $965.94
|
|
|||||
S00123
|
DWK Life Sciences
58014100 |
100 mL |
Pack of 6 Each for $51.64
Case of 2 Pack for $103.28
|
|
|||||
S00125
|
DWK Life Sciences
58014250 |
250 mL |
Pack of 6 Each for $642.00
Case of 2 Pack for $1,284.00
|
|
|||||
S00124
|
DWK Life Sciences
58014200 |
200 mL |
Pack of 6 Each for $595.31
Case of 2 Pack for $1,190.62
|
|
|||||
S00126
|
DWK Life Sciences
58014500 |
500 mL |
Pack of 6 Each for $797.38
Case of 2 Pack for $1,594.76
|
|
|||||
Description
- Kimax™ KG-33 glass Single graduation ring blasted on the neck
- With plug stopcock ground glass stopper and marking spot
Compliance
Glass meets Type 1, Class A, ASTM™ E-438 standards. Designed to ASTM Specification E288, Class A unserialized requirements.
Specifications
Calibrated To Contain | |
Glass | |
Glass |