DWK Life Sciences Kimble™ ValueWare™ Volumetric Flasks, Class A
Calibrated “To Contain.”
Supplier: DWK Life Sciences 58014100
Description
- Kimax™ KG-33 glass Single graduation ring blasted on the neck
- With plug stopcock ground glass stopper and marking spot
Compliance
Glass meets Type 1, Class A, ASTM™ E-438 standards. Designed to ASTM Specification E288, Class A unserialized requirements.
Specifications
Calibrated To Contain | |
100 mL | |
13 |
Glass | |
Glass |