DWK Life Sciences Kimble™ ValueWare™ Volumetric Flasks, Class A, with Snap Cap
Calibrated To Contain.
$179.72 - $1,027.91
Specifications
Closure Material Polyethylene
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
S00134
View Documents
DWK Life Sciences
580082000
2000 mL N/A
S00130
View Documents
DWK Life Sciences
58008100
100 mL N/A
S00132
View Documents
DWK Life Sciences
58008500
500 mL N/A
Description
Description
  • Kimax™ KG-33 glass Class A accuracy but without a serial number
  • With polyethylene snap cap
Compliance
Glass meets Type 1, Class A, ASTM™ E-438 standards. Designed to ASTM Specification E288, Class A unserialized requirements.
Specifications
Specifications
Polyethylene
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'