DWK Life Sciences Kimble™ ValueWare™ Volumetric Flasks, Class A, with Snap Cap
Calibrated To Contain.
Supplier: DWK Life Sciences 58008100
Description
- Kimax™ KG-33 glass Class A accuracy but without a serial number
- With polyethylene snap cap
Compliance
Glass meets Type 1, Class A, ASTM™ E-438 standards. Designed to ASTM Specification E288, Class A unserialized requirements.
Specifications
Polyethylene | |
6.29 in. | |
±0.08 mL |
100 mL | |
16 cm |