missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanadyl(IV) acetylacetonate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 199590500

 View more versions of this product

Catalog No. AC199590500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Vanadyl(IV) acetylacetonate
3153-26-2
Blue-Green to Green
Authentic
Glass bottle
(CH3COCH=C(CH3)O)2V=O
50 g
vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide
JFHJZWAQYMGNBE-SUKNRPLKSA-L
(Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate
131674261
99%
99%
256.0°C to 259.0°C
79°C
98.5% min
C10H14O5V
MFCD00000032
02,456; 03,331; 11,47; 13,54; 15,57; 15,186
Solubility in water: negligible. Other solubilities: soluble in methanol (6.4 g/100 mL), ethanol, ether, chloroform, acetone, dioxane, benzene (0.9 g/100 m, l), pyridine
O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
265.16
265.15
Crystalline Powder

Chemical Identifiers

3153-26-2
265.16
JFHJZWAQYMGNBE-SUKNRPLKSA-L
131674261
O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
C10H14O5V
MFCD00000032
vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide
(Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
If eye irritation persists: Get medical

GHS Signal Word: Warning

EINECSNumber : 221-590-8

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only