missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Vanillin, 99%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 140820010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Vanillin | |
81°C to 84°C | |
1.0600g/mL | |
99% | |
C8H8O3 | |
1 kg | |
15, 10120 | |
MWOOGOJBHIARFG-UHFFFAOYSA-N | |
4-hydroxy-3-methoxybenzaldehyde | |
1183 | |
152.15 | |
Pure |
121-33-5 | |
White to Yellow | |
Authentic | |
Plastic bottle | |
MFCD00006942,MFCD08702848 | |
08, 247 | |
vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde | |
COC1=CC(C=O)=CC=C1O | |
152.15 | |
CHEBI:18346 | |
≥98.5% | |
Crystalline Powder |
Chemical Identifiers
121-33-5 | |
152.15 | |
MWOOGOJBHIARFG-UHFFFAOYSA-N | |
1183 | |
4-hydroxy-3-methoxybenzaldehyde |
C8H8O3 | |
MFCD00006942,MFCD08702848 | |
vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde | |
CHEBI:18346 | |
COC1=CC(C=O)=CC=C1O |
Safety and Handling
Warning
RUO – Research Use Only