missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanillin, 99%, pure, Thermo Scientific Chemicals

$70.60 - $1545.56

Chemical Identifiers

CAS 121-33-5
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00006942,MFCD08702848
InChI Key MWOOGOJBHIARFG-UHFFFAOYSA-N
Synonym vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
PubChem CID 1183
ChEBI CHEBI:18346
IUPAC Name 4-hydroxy-3-methoxybenzaldehyde
SMILES COC1=CC(C=O)=CC=C1O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC140820020
SDS View Documents
Thermo Scientific Chemicals
140820020
2 g Glass Bottle
Each for $70.60
 
AC140821000
SDS View Documents
Thermo Scientific Chemicals
140821000
100 g Plastic bottle
Each for $112.03
 
AC140822500
SDS View Documents
Thermo Scientific Chemicals
140822500
250 g Plastic bottle
Each for $145.75
 
AC140820010
SDS View Documents
Thermo Scientific Chemicals
140820010
1 kg Plastic bottle
Each for $413.46
 
AC140820050
SDS View Documents
Thermo Scientific Chemicals
140820050
5 kg Plastic drum
Each for $1,545.56
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

121-33-5
152.15
MWOOGOJBHIARFG-UHFFFAOYSA-N
1183
4-hydroxy-3-methoxybenzaldehyde
C8H8O3
MFCD00006942,MFCD08702848
vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
CHEBI:18346
COC1=CC(C=O)=CC=C1O

Specifications

121-33-5
White to Yellow
170°C (15.0 mmHg)
Authentic
Glass Bottle
MFCD00006942,MFCD08702848
08, 247
vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
MWOOGOJBHIARFG-UHFFFAOYSA-N
4-hydroxy-3-methoxybenzaldehyde
1183
152.15
Pure
Vanillin
81°C to 84°C
1.0600g/mL
147°C
99%
C8H8O3
2 g
15, 10120
Solubility in water: 10g/L (25°C). Other solubilities: freely soluble in glacial acetic acid,freely soluble in alcohol,chloroform,ether,,carbon disulfide and methanol,soluble in oils,aq. solutions of alkali hydroxide,s
COC1=CC(C=O)=CC=C1O
152.15
CHEBI:18346
99%
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 204-465-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only