missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanillin, 99%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 140820050

 View more versions of this product

Catalog No. AC140820050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Vanillin
81.0°C to 84.0°C
1.0600g/mL
147°C
99%
C8H8O3
5 kg
15, 10120
Solubility in water: 10g/L (25°C). Other solubilities: freely soluble in glacial acetic acid,freely soluble in alcohol,chloroform,ether,,carbon disulfide and methanol,soluble in oils,aq. solutions of alkali hydroxide,s
COC1=CC(C=O)=CC=C1O
152.15
CHEBI:18346
99%
Crystalline Powder
121-33-5
White to Yellow
170.0°C (15.0 mmHg)
Authentic
Plastic drum
MFCD00006942,MFCD08702848
08, 247
vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
MWOOGOJBHIARFG-UHFFFAOYSA-N
4-hydroxy-3-methoxybenzaldehyde
1183
152.15
Pure

Chemical Identifiers

121-33-5
152.15
MWOOGOJBHIARFG-UHFFFAOYSA-N
1183
4-hydroxy-3-methoxybenzaldehyde
C8H8O3
MFCD00006942,MFCD08702848
vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
CHEBI:18346
COC1=CC(C=O)=CC=C1O
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 204-465-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only