missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanillin, 99%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 140821000

 View more versions of this product

Catalog No. AC140821000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Vanillin
81°C to 84°C
1.0600g/mL
99%
C8H8O3
100 g
15, 10120
MWOOGOJBHIARFG-UHFFFAOYSA-N
4-hydroxy-3-methoxybenzaldehyde
1183
152.15
Pure
121-33-5
White to Yellow
Authentic
Plastic bottle
MFCD00006942,MFCD08702848
08, 247
vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
COC1=CC(C=O)=CC=C1O
152.15
CHEBI:18346
≥98.5%
Crystalline Powder

Chemical Identifiers

121-33-5
152.15
MWOOGOJBHIARFG-UHFFFAOYSA-N
1183
4-hydroxy-3-methoxybenzaldehyde
C8H8O3
MFCD00006942,MFCD08702848
vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
CHEBI:18346
COC1=CC(C=O)=CC=C1O
Videos
Safety and Handling

Safety and Handling

Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only