missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Versaflo™ M-400 Series Respiratory Helmet Assembly
Used with an approved air source, M-400 Series Helmets provide respiratory (OSHA APF=1000) and limited head (ANSI Z89.1-2003 Type 1 Class G) and eye (ANSI Z87.1-2010) protection.
Supplier: 3M™ 7100009447
Description
Features:
- Chemical and Scratch Resistant Lenses
- Large Viewing Area
- Lightweight, Easy Visor Replacement System
Environmental Conditions:
- Cold Working Environment
- High Mobility Required
- Safety Helmet Required
Hazard Type:
- Gas, Vapour and Particulates
- Liquid Splash
- Low Energy Flying Particles/Dust
- Falling Objects
- Sparks and Hot Particles
Includes:
- The M-405 assembly includes a M-925 Visor, M-444 Inner Collar, and M-445 Outer Shroud.
- The M-925 visor is made of polycarbonate.
- The M-445 shroud is appropriate for general purpose use.
- The M-407 Assembly includes a M-927 Premium Visor, M-444 Inner Collar, and M-447 Flame Resistant Outer Shroud.
- The M-927 visor is made of coated polycarbonate.
- The M-447 shroud is made of Nomex flame resistant fibers.
- 3M Catalog# M-405 , 3M Catalog# M-407
Specifications
| 1,000 | |
| Black | |
| Demolition, Metalworking, Metal Processing, Smelting, Foundries (Grinding, Polishing), Woodworking, Construction | |
| Standard Shroud, Standard Visor | |
| Non-sterile | |
| Standard | |
| 346 x 346 x 254 mm | |
| 346 mm | |
| 346 mm | |
| 254 mm | |
| 1066 g |
| CE Approved, EN 12941 TH3, EN 14594 3B, EN 166:2001, EN 397 | |
| Respirator Helmet | |
| 3M Air Delivery Units | |
| M-405 | |
| Textile | |
| No | |
| 13.6 in. | |
| 13.6 in. | |
| 9.6 in. | |
| 2.4 lb. |