missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3M™ Versaflo™ M-400 Series Respiratory Helmet Assembly

Used with an approved air source, M-400 Series Helmets provide respiratory (OSHA APF=1000) and limited head (ANSI Z89.1-2003 Type 1 Class G) and eye (ANSI Z87.1-2010) protection.

$1014.23 - $1245.68

Specifications

Assigned Protection Factor (APF) 1,000
Certifications/Compliance CE Approved, EN 12941 TH3, EN 14594 3B, EN 166:2001, EN 397
Color Black
Product Type Respirator Helmet
For Use With (Application) Demolition, Metalworking, Metal Processing, Smelting, Foundries (Grinding, Polishing), Woodworking, Construction
View More Specs

Products 2
Catalog Number Mfr. No. Model Includes Price Quantity  
Catalog Number Mfr. No. Model Includes Price Quantity  
19-039-521
View Documents
3M™
7100009447
M-405 Standard Shroud, Standard Visor
Case for $1,014.23
 
19-039-523
View Documents
3M™
7000002395
M-407 Flame-Resistant Shroud
Case for $1,245.68
 
Description

Description

Features:

  • Chemical and Scratch Resistant Lenses
  • Large Viewing Area
  • Lightweight, Easy Visor Replacement System

Environmental Conditions:

  • Cold Working Environment
  • High Mobility Required
  • Safety Helmet Required

Hazard Type:

  • Gas, Vapour and Particulates
  • Liquid Splash
  • Low Energy Flying Particles/Dust
  • Falling Objects
  • Sparks and Hot Particles

Includes:

  • The M-405 assembly includes a M-925 Visor, M-444 Inner Collar, and M-445 Outer Shroud.
    • The M-925 visor is made of polycarbonate.
    • The M-445 shroud is appropriate for general purpose use.
  • The M-407 Assembly includes a M-927 Premium Visor, M-444 Inner Collar, and M-447 Flame Resistant Outer Shroud.
    • The M-927 visor is made of coated polycarbonate.
    • The M-447 shroud is made of Nomex flame resistant fibers.
  • 3M Catalog# M-405 , 3M Catalog# M-407
Specifications

Specifications

1,000
Black
Demolition, Metalworking, Metal Processing, Smelting, Foundries (Grinding, Polishing), Woodworking, Construction
Non-sterile
No
13.6 in.
13.6 in.
9.6 in.
2.4 lb.
CE Approved, EN 12941 TH3, EN 14594 3B, EN 166:2001, EN 397
Respirator Helmet
3M Air Delivery Units
Textile
346 x 346 x 254 mm
346 mm
346 mm
254 mm
1066 g
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'