missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SP Hotpack™ Vertical Stacking Glassware Washer

Designed for ease of operation and flexible loading, Hotpack Vertical SpaceSaver™ Laboratory Glassware Washers provide large load capacity and built-in flexibility, all in a small footprint.

Supplier:  SP Hotpack™ VSWHA00001

 View more versions of this product

Catalog No. 15-365-000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Equipped with premium features including a soft touch controller, enhanced drying, and quiet run cycles, SP Hotpack Laboratory Glassware Washers are engineered for durability and energy efficient operation. They provide reliable day-in, day-out performance so your laboratory glass is always ready-to-go.

  • Two independently operated chambers with up to four loading levels for high volume washing
  • All models come with removable top and bottom stainless-steel open racks to accommodate a variety of glassware
  • Spindle rack ready levels accommodate direct injection spindle racks (sold separately) for thorough cleaning inside narrow-neck glassware
  • Stainless steel baskets, insert racks and accessories sold separately to maximize space, improve efficiency, and protect specialty glassware
  • Temperature selection to 70°C (158°F); can effectively dissolve stubborn oils
  • DI rinse models ensure a spotless finish; easily connect to fixed or portable DI water systems
  • Optional DI Booster Pump recommended when the incoming DI water source has a pressure of less than 18 psi
  • Delayed start feature allows washer to operate unattended during off-peak hours to reduce energy costs
  • Designed for optimal quiet operation with noise buffering insulation
  • Three filters protect pumps from debris; conveniently located for easy access
  • Minimal, efficient water consumption
  • Enhanced dual fan drying system for spotless results
  • 304 stainless steel chamber and exterior casing ensures corrosion protection and long life
  • 3 in. lockable casters allow easy, controlled mobility of unit
  • UL 61010-1 & CSA 22.2 61010-1 certified by MET Labs
  • Microprocessor control system features 6 cycle selections including a customizable timed option to ensure minimum energy consumption. All wash cycles have pre wash, wash, pre-rinse and rinse cycles to prevent contamination.
  • Rinse Only - Short tap water rinse cycle
  • Normal Wash - Lower temperature (55°C) longer cycle with three (3) final rinse cycles
  • Daily Wash - Standard temperature (65°C) with two (2) final rinse cycles
  • Heavy Wash - Higher temperature (70°C) longer cycle with multiple pre-wash and final rinse cycles
  • Heavy Wash Time Saver - Lower temperature (55°C) version of Heavy Wash with a shorter overall cycle time
  • Timed Wash Programs - Standard temperature (65°C) in programmable 15 minute increments up to 2.5 hours
Specifications

Specifications

Tap Water Rinse - Top and Bottom Standard Racks
UL 61010-1; CSA22.2 61010-1
23.31 in.
120 V
Stainless Steel
Washing Lab Glassware
592 mm
60 Hz
Videos
SDS
Documents

Documents

Promotions

Promotions