missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
SP Hotpack™ Vertical Stacking Glassware Washer
Designed for ease of operation and flexible loading, Hotpack Vertical SpaceSaver™ Laboratory Glassware Washers provide large load capacity and built-in flexibility, all in a small footprint.
$66314.43 - $91122.54
Specifications
Material | Stainless Steel |
---|---|
Certifications/Compliance | UL 61010-1; CSA22.2 61010-1 |
For Use With (Application) | Washing Lab Glassware |
Depth (English) | 23.31 in. |
Depth (Metric) | 592 mm |
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
15-365-000
|
SP Hotpack™
VSWHA00001 |
Tap Water Rinse - Top and Bottom Standard Racks |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15365003
|
SP Hotpack™
VSWHA22001 |
DI Rinse - Top and Bottom Spindle Rack Ready |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Equipped with premium features including a soft touch controller, enhanced drying, and quiet run cycles, SP Hotpack Laboratory Glassware Washers are engineered for durability and energy efficient operation. They provide reliable day-in, day-out performance so your laboratory glass is always ready-to-go.
- Two independently operated chambers with up to four loading levels for high volume washing
- All models come with removable top and bottom stainless-steel open racks to accommodate a variety of glassware
- Spindle rack ready levels accommodate direct injection spindle racks (sold separately) for thorough cleaning inside narrow-neck glassware
- Stainless steel baskets, insert racks and accessories sold separately to maximize space, improve efficiency, and protect specialty glassware
- Temperature selection to 70°C (158°F); can effectively dissolve stubborn oils
- DI rinse models ensure a spotless finish; easily connect to fixed or portable DI water systems
- Optional DI Booster Pump recommended when the incoming DI water source has a pressure of less than 18 psi
- Delayed start feature allows washer to operate unattended during off-peak hours to reduce energy costs
- Designed for optimal quiet operation with noise buffering insulation
- Three filters protect pumps from debris; conveniently located for easy access
- Minimal, efficient water consumption
- Enhanced dual fan drying system for spotless results
- 304 stainless steel chamber and exterior casing ensures corrosion protection and long life
- 3 in. lockable casters allow easy, controlled mobility of unit
- UL 61010-1 & CSA 22.2 61010-1 certified by MET Labs
- Microprocessor control system features 6 cycle selections including a customizable timed option to ensure minimum energy consumption. All wash cycles have pre wash, wash, pre-rinse and rinse cycles to prevent contamination.
- Rinse Only - Short tap water rinse cycle
- Normal Wash - Lower temperature (55°C) longer cycle with three (3) final rinse cycles
- Daily Wash - Standard temperature (65°C) with two (2) final rinse cycles
- Heavy Wash - Higher temperature (70°C) longer cycle with multiple pre-wash and final rinse cycles
- Heavy Wash Time Saver - Lower temperature (55°C) version of Heavy Wash with a shorter overall cycle time
- Timed Wash Programs - Standard temperature (65°C) in programmable 15 minute increments up to 2.5 hours
Specifications
Stainless Steel | |
Washing Lab Glassware | |
592 mm | |
60 Hz |
UL 61010-1; CSA22.2 61010-1 | |
23.31 in. | |
120 V |