missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Vikan™ 10 in. Double Blade Ultra Hygiene Squeegee
Vikan™ 10 in. Double Blade Ultra Hygiene Squeegee features double rubber blades that are non-porous and easy to clean.
Supplier: Vikan™ 77113
Description
- Ideal for drying surfaces after a sanitizing process or for pushing food from one area to another
- Effective in dry production areas
- Double rubber blades are non-porous and easy to clean
- Cartridge system blades can be removed for easy cleaning and replaced when worn out
- Economical hygienic option
- Color-coded
Specifications
FDA Compliant | |
Thread type Euro Hygienic | |
1 Ea. | |
3.5 in. |
Blue | |
Polypropylene, Rubber | |
Small |