missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vinyl bromide, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 428771000

 View more versions of this product

Catalog No. AC428771000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Vinyl bromide
593-60-2,109-99-9
−17°C
C2H3Br
MFCD00000183
0.927
INLLPKCGLOXCIV-UHFFFAOYSA-N
bromoethene
11641
106.95
1M solution in THF
0.9270g/mL
AcroSeal™ Glass Bottle
H2C=CHBr
100 mL
vinyl bromide, bromoethylene, ethene, bromo, monobromoethylene, vinylbromid, bromure de vinyle, vinylbromide, polyvinyl bromide, ethylene, bromo, polyvinylbromide
BrC=C
106.95
CHEBI:51311
Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only