VistaLab™ Technologies Ovation™ Fixed-Volume Pipetter
Ovation™ pipettes have a unique and patented ergonomic design. They are the only pipettes that fully address the issues of posture, repetition, and force to significantly reduce strain and pain that occur from pipetting, while always providing superior accuracy and precision.
$545.71 - $2636.49
Specifications
Autoclavable | Not Autoclavable |
---|---|
Number of Channels | 1 |
Sterility | Non-sterile |
For Use With (Application) | Micro Pipetting/Liquid Transfer |
Catalog Number | Mfr. No. | Accuracy | Volume (Metric) | Type | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Accuracy | Volume (Metric) | Type | Price | Quantity | |||
22-146-607
|
VistaLab™ Technologies
11570010 |
±1.5% | 10 μL | Single Stroke Pipetter |
Each for $610.10
|
|
|||
22-146-608
|
VistaLab™ Technologies
11570020 |
±1.0% | 20 μL | Single Stroke Pipetter |
Each for $609.10
|
|
|||
22-146-609
|
VistaLab™ Technologies
11570025 |
±1.4% | 25 μL | Single Stroke Pipetter |
Each for $545.71
|
|
|||
22-146-610
|
VistaLab™ Technologies
11570050 |
±0.8% | 50 μL | Single Stroke Pipetter |
Each for $545.71
|
|
|||
22-146-611
|
VistaLab™ Technologies
11570100 |
±0.8% | 100 μL | Single Stroke Pipetter |
Each for $609.10
|
|
|||
22-146-612
|
VistaLab™ Technologies
11570200 |
±0.8% | 200 μL | Single Stroke Pipetter |
Each for $609.10
|
|
|||
22-146-615
|
VistaLab™ Technologies
11570500 |
±0.8% | 500 μL | Single Stroke Pipetter |
Each for $609.10
|
|
|||
22-146-616
|
VistaLab™ Technologies
11571000 |
±0.8% | 1000 μL | Single Stroke Pipetter |
Each for $609.10
|
|
|||
22-146-690
|
VistaLab™ Technologies
11570999 |
±1.5%/ ±0.8%/ ±0.8%/ ±0.8% | 10, 100, 200, 1000 μL | Fixed Volume Pipette |
Pack of 6 Each for $2,636.49
|
|
|||
22-146-618
|
VistaLab™ Technologies
12570002 |
±1.5% | 2 μL | Two Stroke Pipette |
Each for $621.33
|
|
|||
22-146-623
|
VistaLab™ Technologies
12570050 |
±0.8% | 50 μL | Two Stroke Pipette |
Each for $609.10
|
|
|||
22-146-629
|
VistaLab™ Technologies
12571000 |
±0.8% | 1000 μL | Two Stroke Pipette |
Each for $609.10
|
|
|||
Description
- Mechanical 1-stroke (F1) or 2-stroke (with overblow) (F2) low-force plunger operation
- Ovation F1 available from 5-1000 μL
- Ovation F2 available from 1-1000 μL
- Custom fixed volumes available upon request
- Built in microtube cap opener
- Nozzle with replaceable filter prevents contamination of pipetter
- In-lab calibration adjustment possible
Specifications
Not Autoclavable | |
1 | |
Non-sterile | |
Micro Pipetting/Liquid Transfer |