missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Vitamin E, 1 mg = 1.1 IU, USP, 96-102%, Spectrum™ Chemical
VI135, 10191-41-0, C29H50O2
Supplier: Spectrum Chemical Mfg Cor VI1351LTGL
Description
Spectrum™ Chemical Vitamin E, 1 mg = 1.1 IU, USP is used as an antioxidant. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
1.0 mL | |
1 | |
C29H50O2 | |
1 L | |
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1 | |
430.72 | |
USP |
10191-41-0 | |
Amber Glass Bottle | |
MFCD00072051 | |
GVJHHUAWPYXKBD-IEOSBIPESA-N | |
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol | |
96 to 102% |
Chemical Identifiers
10191-41-0 | |
430.72 | |
GVJHHUAWPYXKBD-IEOSBIPESA-N | |
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1 |
C29H50O2 | |
MFCD00072051 | |
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol |