missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Vitamin E, 1 mg = 1.1 IU, USP, 96-102%, Spectrum™ Chemical
VI135, 10191-41-0, C29H50O2
$422.00 - $24472.85
Chemical Identifiers
CAS | 10191-41-0 |
---|---|
Molecular Formula | C29H50O2 |
Molecular Weight (g/mol) | 430.72 |
MDL Number | MFCD00072051 |
InChI Key | GVJHHUAWPYXKBD-IEOSBIPESA-N |
IUPAC Name | (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol |
SMILES | CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18607725
|
Spectrum Chemical Mfg Cor
VI135100ML |
100 mL | Amber Glass Bottle |
Each for $422.00
|
|
||||
18607728
|
Spectrum Chemical Mfg Cor
VI135500ML |
500 mL | Amber Glass Bottle |
Each for $1,301.21
|
|
||||
18607726
|
Spectrum Chemical Mfg Cor
VI1351LTGL |
1 L | Amber Glass Bottle |
Each for $2,590.21
|
|
||||
18607727
|
Spectrum Chemical Mfg Cor
VI13520KGBL |
20 kg | Steel Pail |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Vitamin E, 1 mg = 1.1 IU, USP is used as an antioxidant. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
10191-41-0 | |
430.72 | |
GVJHHUAWPYXKBD-IEOSBIPESA-N | |
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1 |
C29H50O2 | |
MFCD00072051 | |
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol |
Specifications
1.0 mL | |
1 | |
C29H50O2 | |
100 mL | |
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1 | |
430.72 | |
USP |
10191-41-0 | |
Amber Glass Bottle | |
MFCD00072051 | |
GVJHHUAWPYXKBD-IEOSBIPESA-N | |
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol | |
96 to 102% |