missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
VITLAB™ VITsafe™ Wide-Mouth Wash Bottle
VITLAB™ VITsafe™ Wide-Mouth Wash Bottle with integrated vent in cap to reduce dripping.
Supplier: VITLAB™ 1352909
Description
- Chemical name in German, English, French and Spanish
- Chemical formula
- CAS Number, hazard pictogram, signal word
- Risk Phrases (H phrases), safety phrases (P phrases)
- NFPA Code
- Fine tip polypropylene tube provides directional accuracy
- Colored cap allows easier identification
Specifications
500 mL | |
16.9 oz. | |
i-Hexane | |
166 mm | |
6 | |
76 mm | |
Not Autoclavable | |
No | |
Cylindrical | |
Safety-labeled Wash Bottle | |
6.54 in. | |
i-Hexane | |
76 mm | |
Vented Tube |
500 mL | |
Red | |
LDPE, Polypropylene | |
Pack | |
Non-sterile | |
Yes | |
Reusable | |
Wide, Threaded | |
166 mm | |
3 in. | |
6.54 in. | |
GL 45 | |
Spouted Cap |