missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
VITLAB™ VITsafe™ Wide-Mouth Wash Bottle
VITLAB™ VITsafe™ Wide-Mouth Wash Bottle with integrated vent in cap to reduce dripping.
$368.21 - $707.85
Specifications
Capacity | 500 mL |
---|---|
Capacity (Metric) | 500 mL |
Capacity (English) | 16.9 oz. |
Length (Metric) | 166 mm |
Packaging | Pack |
Catalog Number | Mfr. No. | Chemical Label | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Chemical Label | Price | Quantity | |||||
14-380-960
|
VITLAB™
1352869 |
Ethanol |
Pack of 12 Each for $707.85
|
|
|||||
14-380-956
|
VITLAB™
1352899 |
Heptane |
Pack of 6 Each for $368.21
|
|
|||||
14-380-957
|
VITLAB™
1352909 |
i-Hexane |
Pack of 6 Each for $368.21
|
|
|||||
14-380-959
|
VITLAB™
1352819 |
Distilled Water |
Pack of 12 Each for $707.85
|
|
|||||
14-380-958
|
VITLAB™
1352959 |
Xylene |
Pack of 6 Each for $368.21
|
|
|||||
Description
- Chemical name in German, English, French and Spanish
- Chemical formula
- CAS Number, hazard pictogram, signal word
- Risk Phrases (H phrases), safety phrases (P phrases)
- NFPA Code
- Fine tip polypropylene tube provides directional accuracy
- Colored cap allows easier identification
Specifications
500 mL | |
16.9 oz. | |
Pack | |
Yes | |
Reusable | |
Cylindrical | |
Safety-labeled Wash Bottle | |
GL 45 | |
Vented Tube |
500 mL | |
166 mm | |
Non-sterile | |
Not Autoclavable | |
No | |
166 mm | |
6.54 in. | |
Spouted Cap |