W.S. TYLER™ RO-TAP™ Sieve Shakers
Recommended for accurate and consistent particle analysis testing
Supplier:  W.S. TYLER™ RX29
 View more versions of this product
Catalog No. 04-909
Explore available promotions
Description
Description
Recommended for accurate and consistent particle analysis testing
The shaker has a unique two dimentional operation: a horizontal, circular motion and a vertical, topping motion which allow material particles to stratify and seek critical openings in the test media.
  • Heavy duty, stationary unit With digital 99 min. timer/clock, accurate to 0.1 second
  • Requires minimal maintenance
  • Each unit test run and serialized before shipment
  • Sieves are sold separately
Specifications
Specifications
110 V, 60 Hz
5.4 A
60 Hz
8 in. (Sieve)
99 min.
110 V
Sieve Shaker
8 in.
Accommodates 6 full-height sieves or 13 half-height sieves
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories