W.S. TYLER™ RO-TAP™ Sieve Shakers
Recommended for accurate and consistent particle analysis testing
Supplier:  W.S. TYLER™ RX30
 View more versions of this product
Catalog No. 04-909-3
Explore available promotions
Description
Description
Recommended for accurate and consistent particle analysis testing
The shaker has a unique two dimentional operation: a horizontal, circular motion and a vertical, topping motion which allow material particles to stratify and seek critical openings in the test media.
  • Heavy duty, stationary unit With digital 99 min. timer/clock, accurate to 0.1 second
  • Requires minimal maintenance
  • Each unit test run and serialized before shipment
  • Sieves are sold separately
Specifications
Specifications
110 V, 60 Hz
Sieve Shaker
21 in.
12 in.
64 cm
70 cm
99 min.
110 V
60 Hz
540 cm
25.1 in.
28 in.
Accommodates 4 full-height sieves (w/ FH pan), 6 full-hgt. sieves (w/ HH pan); or 8 half-hgt. sieves (w/ HH pan)