missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Poltex Wall Pipette Rack, 5 Positions

PETG constructed pipette hanger with 5 positions.

$337.94 - $414.76

Specifications

Product Type Pipette Rack
Array 1 x 5
Autoclavable Not Autoclavable
Color Clear
Holds Pipettes
View More Specs

Products 3
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
22000752 DFS Item
View Documents
Poltex
PIPRKWL5M
Magnet Mount
Each for $414.76
 
22000753
View Documents
Poltex
PIPRKWL5T
VHB Tape
Each for $404.53
 
22000754
View Documents
Poltex
PIPRKWL5W
Wall Mount
Each for $337.94
 
Description

Description

  • Fits most standard brands of Pipettes
  • Choose from wall mount, powerful magnets, or VHB tape
Specifications

Specifications

Pipette Rack
Not Autoclavable
Pipettes
5
None
Non-sterile
1 x 5
Clear
PETG
Rectangular, Rounded Corners
No
9 x 4 x 5 in.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'