missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Conair™ Waring™ Laboratory Blenders: Two Speeds, Commercial
Commercial-grade units for demanding mixing and blending applications
$976.87 - $976.87
Specifications
Amperage | 3.6 A |
---|---|
Speed | 18,000, 22,000 rpm |
Hertz | 50/60 Hz |
Horsepower | 0.4 hp |
Base Material | Epoxy-coated Metal |
Catalog Number | Mfr. No. | Dimensions (L x W x H) | Voltage | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W x H) | Voltage | Price | Quantity | ||||
14-509-17
|
Conair™
WRG7011S |
8.5 x 7.25 x 15 in. (21.59 x 18.41 x 38 cm) | 115/120 V |
Each for $976.87
|
|
||||
Description
Commercial-grade units for demanding mixing and blending applications
- Cloverleaf design
- Choice of glass and/or stainless-steel containers
- Sturdy, diecast metal bases and stainless-steel blades
- All durably constructed for years of dependable service
- Epoxy-coated bases with chrome top pieces on all models, except for the aluminum and steel base (Hazardous-Location Model)
- Heat-resistant borosilicate glass (integral handle) or stainless-steel (attached handles)
- Two-piece vinyl and styrene lids on all models
Order Info
Electrical requirements for all models: 115/120V 50/60Hz
Certifications
UL listed (Select models only; see ordering chart)
Specifications
3.6 A | |
50/60 Hz | |
Epoxy-coated Metal | |
1 Ea. |
18,000, 22,000 rpm | |
0.4 hp | |
Base, container, 3-wire grounded cord and plug |