Conair™ Waring™ Mini-sample Containers for Blender
Stainless steel containers
Supplier:  Conair™ MC2
 View more versions of this product
Catalog No. 14-509-18B
Explore available promotions
Description
Description
Stainless-steel blending assemblies are for use with 1L (34 oz.) capacity blenders (nominal capacity; actual capacity depends on container used).
  • Snap-fit cover
  • Cloverleaf design
Specifications
Specifications
Mini Sample Container
37 to 110 mL
Waring™ 1 L blender
Product Suggestions
Product Suggestions