missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ Warning Labels, Pictogram: Battery Hazard

Feature pictograms that indicate battery hazard. Brady™ Warning Labels will adhere to any clean, dry surface and are available in a variety of sizes.

Supplier:  Brady™ 60219

 View more versions of this product

Catalog No. 19-107-277



Description

Description

  • Made with highly visible B-946 bright colored vinyl
  • Adheres to any clean, dry surface
  • Available in 5 different sizes
Specifications

Specifications

Pictogram: Battery Hazard
Black, Yellow
Warning Label
Triangle
0.004 in.
Acrylic, Permanent, Cold-temperature
Pressure-sensitive Vinyl (B-946)
Indoor/Outdoor
4 x 4 in.
SDS
Documents

Documents

Promotions

Promotions