missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Warning Labels, Pictogram: Battery Hazard
Feature pictograms that indicate battery hazard. Brady™ Warning Labels will adhere to any clean, dry surface and are available in a variety of sizes.
$100.47 - $207.09
Specifications
Description | Pictogram: Battery Hazard |
---|---|
Adhesive Type | Acrylic, Permanent, Cold-temperature |
Color | Black, Yellow |
Material | Pressure-sensitive Vinyl (B-946) |
Product Type | Warning Label |
Catalog Number | Mfr. No. | Size (English) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size (English) | Price | Quantity | |||||
19-107-256
|
Brady™
60186 |
0.5 x 0.5 in. |
Pack of 10 Each for $100.47
|
|
|||||
19-107-263
|
Brady™
60197 |
1 x 1 in. |
Pack of 10 Each for $105.58
|
|
|||||
19-107-270
|
Brady™
60208 |
2 x 2 in. |
Pack of 10 Each for $183.72
|
|
|||||
19-107-277
|
Brady™
60219 |
4 x 4 in. |
Pack of 10 Each for $123.84
|
|
|||||
19-107-284
|
Brady™
BRD60230 |
8 x 8 in. |
Pack of 10 Each for $207.09
|
|
|||||
Description
- Made with highly visible B-946 bright colored vinyl
- Adheres to any clean, dry surface
- Available in 5 different sizes
Specifications
Pictogram: Battery Hazard | |
Black, Yellow | |
Warning Label | |
Triangle |
Acrylic, Permanent, Cold-temperature | |
Pressure-sensitive Vinyl (B-946) | |
Indoor/Outdoor | |
0.004 in. |