missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ Warning Labels, Pictogram: Explosive Material

Feature pictograms that indicate hazards for explosive material. Brady™ Warning Labels will adhere to any clean, dry surface and are available in a variety of sizes.

Supplier:  Brady™ BRD60215

 View more versions of this product

Catalog No. 19-107-274


Explore available promotions

Description

Description

  • Made with highly visible B-946 bright colored vinyl
  • Adheres to any clean, dry surface
  • Available in 5 different sizes
Specifications

Specifications

Pictogram: Explosive Material
Black, Yellow
Warning Label
Triangle
0.004 in.
Acrylic, Permanent, Cold-temperature
Pressure-sensitive Vinyl (B-946)
Indoor/Outdoor
4 x 4 in.
Videos
SDS
Documents

Documents

Promotions

Promotions