missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Warning Labels, Pictogram: Explosive Material
Feature pictograms that indicate hazards for explosive material. Brady™ Warning Labels will adhere to any clean, dry surface and are available in a variety of sizes.
$95.15 - $200.39
Specifications
Description | Pictogram: Explosive Material |
---|---|
Adhesive Type | Acrylic, Permanent, Cold-temperature |
Color | Black, Yellow |
Material | Pressure-sensitive Vinyl (B-946) |
Product Type | Warning Label |
Catalog Number | Mfr. No. | Size (English) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size (English) | Price | Quantity | |||||
19-107-253
|
Brady™
60182 |
0.5 x 0.5 in. |
Pack of 10 Each for $95.15
|
|
|||||
19-107-260
|
Brady™
60193 |
1 x 1 in. |
Pack of 10 Each for $101.56
|
|
|||||
19-107-267
|
Brady™
60204 |
2 x 2 in. |
Pack of 10 Each for $177.51
|
|
|||||
19-107-274
|
Brady™
BRD60215 |
4 x 4 in. |
Pack of 10 Each for $118.95
|
|
|||||
19-107-281
|
Brady™
60226 |
8 x 8 in. |
Pack of 10 Each for $200.39
|
|
|||||
Description
- Made with highly visible B-946 bright colored vinyl
- Adheres to any clean, dry surface
- Available in 5 different sizes
Specifications
Pictogram: Explosive Material | |
Black, Yellow | |
Warning Label | |
Triangle |
Acrylic, Permanent, Cold-temperature | |
Pressure-sensitive Vinyl (B-946) | |
Indoor/Outdoor | |
0.004 in. |